Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127561.1 | NP_001121033.1 | 490 | 79 | ||
GenBank mRNA | BC160913.1 | - | 490 | 7457160 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus |