Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meis1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134702.1 | NP_001128174.1 | 4 - 5 | 484 | 143 | |
XM_006251529.2 | XP_006251591.1 | 4 - 5 | 1732 | 143 | ||
GenBank mRNA | BC161984.1 | - | 4 - 5 | 484 | 9030600 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |