Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135809.1 | NP_001129281.1 | 2 - 3 | 262 | 90 | |
GenBank mRNA | DQ537495.1 | - | 2 - 3 | 262 | 90 |
Gene Symbol: LOC100359989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233654.2 | XP_006233716.2 | 2 - 3 | 211 | 90 |
Gene Symbol: LOC103691756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761631.1 | XP_008759853.1 | 1 - 2 | 109 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |