Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135899.1 | NP_001129371.1 | 4 - 5 | 647 | 73 | |
GenBank mRNA | BC127545.1 | - | 4 - 5 | 647 | 7321962 | 73 |
FQ211046.1 | - | 4 - 5 | 703 | 73 | ||
FQ219104.1 | - | 4 - 5 | 691 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |