Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022634.2 | NP_072156.2 | 1 - 2 | 37 | 104 | |
XM_006256078.2 | XP_006256140.1 | 1 - 2 | 178 | 104 | ||
XM_006256079.2 | XP_006256141.1 | 1 - 2 | 178 | 104 | ||
GenBank mRNA | AF208230.1 | - | 1 - 2 | 37 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |