Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100494.1 | NP_001093964.1 | 7 - 8 | 876 | 90 | |
XM_006246643.2 | XP_006246705.1 | 8 - 9 | 1013 | 90 | ||
XM_006246644.2 | XP_006246706.1 | 8 - 9 | 1001 | 90 | ||
XM_008767804.1 | XP_008766026.1 | 7 - 8 | 917 | 90 | ||
GenBank mRNA | BC087638.1 | - | 5 - 6 | 510 | 90 | |
BC107474.1 | - | 6 - 7 | 537 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |