Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173428.1 | NP_001166899.1 | 2 - 3 | 737 | 69 | |
XM_006228478.2 | XP_006228540.1 | 2 - 3 | 732 | 69 | ||
XM_006228479.2 | XP_006228541.1 | 2 - 3 | 644 | 69 | ||
XM_008758985.1 | XP_008757207.1 | 2 - 3 | 840 | 69 | ||
XM_008758987.1 | XP_008757209.1 | 2 - 3 | 897 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |