Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053461.1 | NP_445913.1 | 1 | 247 | 84 | |
GenBank mRNA | AF334107.2 | - | 1 | 74 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Both primers and probe map within a single exon |