Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013112.1 | NP_037244.1 | 2 - 3 | 188 | 99 | |
XM_006250238.2 | XP_006250300.1 | 3 - 4 | 322 | 99 | ||
XM_006250239.2 | XP_006250301.1 | 3 - 4 | 328 | 99 | ||
GenBank mRNA | FQ209753.1 | - | 3 - 4 | 253 | 99 | |
FQ210214.1 | - | 3 - 4 | 246 | 99 | ||
FQ210325.1 | - | 3 - 4 | 273 | 99 | ||
FQ210462.1 | - | 3 - 4 | 251 | 99 | ||
FQ210537.1 | - | 2 - 3 | 247 | 99 | ||
FQ210542.1 | - | 2 - 3 | 376 | 99 | ||
FQ210648.1 | - | 3 - 4 | 250 | 99 | ||
FQ210885.1 | - | 3 - 4 | 280 | 99 | ||
FQ210979.1 | - | 3 - 4 | 253 | 99 | ||
FQ211265.1 | - | 3 - 4 | 250 | 99 | ||
FQ211383.1 | - | 3 - 4 | 252 | 99 | ||
FQ218273.1 | - | 3 - 4 | 253 | 99 | ||
FQ218326.1 | - | 3 - 4 | 247 | 99 | ||
FQ218756.1 | - | 3 - 4 | 254 | 99 | ||
FQ221741.1 | - | 3 - 4 | 253 | 99 | ||
M28615.1 | - | 2 - 3 | 157 | 99 | ||
X03468.1 | - | 2 - 3 | 188 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |