Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031578.1 | NP_113766.1 | 5 - 6 | 1734 | 71 | |
GenBank mRNA | BC081773.1 | - | 5 - 6 | 1330 | 7131899 | 71 |
D50864.1 | - | 5 - 6 | 1734 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |