Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031578.1 | NP_113766.1 | 6 - 7 | 1829 | 95 | |
GenBank mRNA | BC081773.1 | - | 6 - 7 | 1425 | 7131899 | 95 |
D50864.1 | - | 6 - 7 | 1829 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |