Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032075.3 | NP_114464.1 | 1 - 2 | 794 | 63 | |
GenBank mRNA | AB001982.1 | - | 1 - 2 | 794 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Rattus norvegicus Probe spans exons |