Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053656.2 | NP_446108.2 | 8 - 9 | 905 | 83 | |
XM_006249515.2 | XP_006249577.1 | 8 - 9 | 1098 | 83 | ||
XM_006249516.2 | XP_006249578.1 | 8 - 9 | 1097 | 83 | ||
XM_006249517.2 | XP_006249579.1 | 8 - 9 | 1060 | 83 | ||
XM_008769305.1 | XP_008767527.1 | 7 - 8 | 981 | 83 | ||
XM_008769306.1 | XP_008767528.1 | 8 - 9 | 774 | 83 | ||
XM_008769307.1 | XP_008767529.1 | 6 - 7 | 846 | 83 | ||
GenBank mRNA | AF013241.1 | - | 1 - 2 | 130 | 83 | |
AF020756.1 | - | 7 - 8 | 805 | 83 | ||
AF020757.1 | - | 7 - 8 | 851 | 83 | ||
AF020758.1 | - | 6 - 7 | 945 | 83 | ||
AF020759.1 | - | 8 - 9 | 867 | 83 | ||
AF064549.1 | - | 7 - 8 | 942 | 83 | ||
U14414.1 | - | 8 - 9 | 905 | 83 | ||
Y10474.1 | - | 8 - 9 | 887 | 83 | ||
Y10475.1 | - | 8 - 9 | 941 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |