Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spetex-2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009976.1 | NP_001009976.1 | 2 - 3 | 296 | 84 | |
GenBank mRNA | AB180076.1 | - | 2 - 3 | 296 | 84 |
Gene Symbol: Spetex-2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011697.1 | NP_001011697.1 | 2 - 3 | 251 | 84 | |
GenBank mRNA | AB180077.1 | - | 2 - 3 | 251 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |