Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rup2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034950.2 | NP_001030122.2 | 1 - 2 | 116 | 89 | |
GenBank mRNA | AF198441.1 | - | 1 - 2 | 88 | 89 | |
AY327506.1 | - | 1 - 2 | 116 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |