Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076793.1 | NP_001070261.1 | 5 - 6 | 712 | 86 | |
XM_006228225.2 | XP_006228287.1 | 4 - 5 | 933 | 86 | ||
XM_008758988.1 | XP_008757210.1 | 5 - 6 | 735 | 86 | ||
XM_008758989.1 | XP_008757211.1 | 4 - 5 | 699 | 86 | ||
GenBank mRNA | AY998481.1 | - | 5 - 6 | 712 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |