Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lilrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100123.1 | NP_001093593.1 | 4 - 5 | 705 | 90 | |
XM_006228169.2 | XP_006228231.1 | 3 - 4 | 752 | 90 | ||
GenBank mRNA | AY998483.1 | - | 4 - 5 | 684 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |