Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl37a-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038113.1 | 147 | 99 | |||
GenBank mRNA | C06947.1 | - | 135 | 99 | ||
X14069.1 | - | 159 | 99 |
Gene Symbol: Rpl37a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108801.1 | NP_001102271.1 | 2 - 3 | 187 | 99 | |
NM_001205318.1 | NP_001192247.1 | 2 - 3 | 157 | 99 | ||
GenBank mRNA | AA685577.1 | - | 1 | 157 | 99 | |
BP477484.1 | - | 2 - 3 | 99 | |||
CB579886.1 | - | 1 - 2 | 433 | 99 | ||
CF108195.1 | - | 1 - 2 | 136 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |