Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012657.2 | NP_036789.2 | 5 | 1257 | 113 | |
XM_006240458.2 | XP_006240520.1 | 5 | 1300 | 113 | ||
GenBank mRNA | BC062236.1 | - | 5 | 6888893 | 113 | |
D00751.1 | - | 5 | 1210 | 113 | ||
FQ209577.1 | - | 5 | 1259 | 113 | ||
FQ209663.1 | - | 4 | 985 | 113 | ||
FQ210286.1 | - | 5 | 1263 | 113 | ||
FQ218085.1 | - | 5 | 1262 | 113 | ||
FQ218647.1 | - | 4 | 934 | 113 | ||
FQ218802.1 | - | 5 | 1259 | 113 | ||
FQ219296.1 | - | 5 | 1263 | 113 | ||
FQ219312.1 | - | 5 | 1261 | 113 | ||
FQ219600.1 | - | 5 | 1257 | 113 | ||
M14320.1 | - | 4 | 113 | |||
M15916.1 | - | 5 | 113 | |||
X05348.1 | - | 5 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |