Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191670.1 | NP_001178599.1 | 6 - 7 | 1772 | 87 | |
XM_006249937.2 | XP_006249999.1 | 7 - 8 | 2001 | 87 | ||
XM_006249938.2 | XP_006250000.1 | 7 - 8 | 1966 | 87 | ||
XM_006249939.2 | XP_006250001.1 | 7 - 8 | 1992 | 87 | ||
XM_006249940.2 | XP_006250002.1 | 7 - 8 | 2044 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |