Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191800.1 | NP_001178729.1 | 1 - 2 | 84 | 85 | |
GenBank mRNA | CK481225.1 | - | 256 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |