Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Siglec15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191942.1 | NP_001178871.1 | 1 - 2 | 52 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |