Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100359907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195414.1 | NP_001182343.1 | 2 - 3 | 166 | 96 | |
GenBank mRNA | HM015624.1 | - | 2 - 3 | 166 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |