Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197217.1 | NP_001184146.1 | 3 - 4 | 517 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |