Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134933.2 | NP_001128405.1 | 5 - 6 | 354 | 101 | |
NM_001244867.1 | NP_001231796.1 | 4 - 5 | 302 | 101 | ||
GenBank mRNA | AI716760.1 | - | 1 - 2 | 101 | ||
CB729209.1 | - | 5 - 6 | 354 | 101 | ||
FQ223262.1 | - | 4 - 5 | 302 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |