Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073140.1 | 2 - 3 | 239 | 84 | ||
XM_008760728.1 | XP_008758950.1 | 2 - 3 | 251 | 84 | ||
GenBank mRNA | BC161924.1 | - | 2 - 3 | 216 | 8370925 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
NADH dehydrogenase (ubiquinone) complex I, assembly factor 2 |
Ndufaf2 |
Ndufa12l, RGD1560158 |
-4.8kb | Anti-sense | Chr. 2 - 39322145 - 39434537 on NCBI Build Rnor_6.0 |
Downstream | DEP domain containing 1B | Depdc1b | 534.1kb | Sense | Chr. 2 - 39980802 - 40069982 on NCBI Build Rnor_6.0 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |