Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004278.3 | NP_001004278.2 | 1 - 2 | 170 | 82 | |
NM_001270736.1 | NP_001257665.1 | 1 - 2 | 170 | 82 | ||
NM_001270737.1 | NP_001257666.1 | 1 - 2 | 170 | 82 | ||
XM_006230847.2 | XP_006230909.1 | 1 - 2 | 445 | 82 | ||
GenBank mRNA | AY690667.1 | - | 1 - 2 | 187 | 82 | |
BC097254.1 | - | 1 - 2 | 151 | 7453704 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |