Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037975.2 | NP_001033064.2 | 17 - 18 | 2142 | 108 | |
XM_006237281.2 | XP_006237343.1 | 17 - 18 | 2154 | 108 | ||
GenBank mRNA | J03552.1 | - | 17 - 18 | 2142 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |