Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128139.2 | NP_001121611.1 | 5 - 6 | 509 | 80 | |
GenBank mRNA | DQ537493.1 | - | 5 - 6 | 511 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |