Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270736.1 | NP_001257665.1 | 4 - 5 | 1145 | 71 | |
XM_006230847.2 | XP_006230909.1 | 4 - 5 | 1420 | 71 | ||
XM_008760155.1 | XP_008758377.1 | 4 - 5 | 1430 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |