Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271054.1 | NP_001257983.1 | 8 - 9 | 1017 | 98 | |
XM_006240151.2 | XP_006240213.1 | 8 - 9 | 1067 | 98 | ||
XM_006240152.2 | XP_006240214.1 | 9 - 10 | 1147 | 98 | ||
XM_006240153.2 | XP_006240215.1 | 8 - 9 | 1491 | 98 | ||
GenBank mRNA | AF441249.1 | - | 7 - 8 | 794 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |