Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271180.1 | NP_001258109.1 | 1 - 2 | 162 | 74 | |
GenBank mRNA | BC088239.1 | - | 1 - 2 | 125 | 74 | |
FQ213935.1 | - | 1 - 2 | 153 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |