Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271229.1 | NP_001258158.1 | 5 - 6 | 917 | 74 | |
GenBank mRNA | BC168162.1 | - | 4 - 5 | 1151 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons |