Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021595.2 | NP_067606.1 | 2 - 3 | 402 | 82 | |
XM_008763255.1 | XP_008761477.1 | 2 - 3 | 912 | 82 | ||
XM_008763256.1 | XP_008761478.1 | 3 - 4 | 829 | 82 | ||
XM_008763257.1 | XP_008761479.1 | 3 - 4 | 588 | 82 | ||
XM_008763258.1 | XP_008761480.1 | 3 - 4 | 739 | 82 | ||
XM_008763259.1 | XP_008761481.1 | 2 - 3 | 548 | 82 | ||
GenBank mRNA | AB040815.1 | - | 2 - 3 | 403 | 82 | |
AF250322.1 | - | 2 - 3 | 403 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |