Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Star | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031558.3 | NP_113746.1 | 2 - 3 | 298 | 61 | |
GenBank mRNA | AB001349.1 | - | 254 | 61 | ||
AB006007.1 | - | 2 - 3 | 299 | 61 | ||
AF044081.1 | - | 238 | 61 | |||
AY736357.1 | - | 2 - 3 | 179 | 61 | ||
BC088859.1 | - | 2 - 3 | 190 | 7311249 | 61 | |
EF091859.1 | - | 1 - 2 | 45 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |