Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053967.2 | NP_446419.1 | 3 - 4 | 1370 | 97 | |
GenBank mRNA | BC081748.1 | - | 3 - 4 | 1311 | 7134081 | 97 |
X89999.1 | - | 3 - 4 | 1374 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |