Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020543.4 | NP_065418.3 | 1 | 847 | 99 | |
GenBank mRNA | AF218846.1 | - | 1 | 847 | 99 | |
FJ694960.1 | - | 2 | 1015 | 99 | ||
FJ694961.1 | - | 2 | 1117 | 99 | ||
JN420349.1 | - | 3 | 1212 | 99 | ||
JX494784.1 | - | 2 | 1117 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Inventoried Both primers and probe map within a single exon |