Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276491.1 | NP_001263420.1 | 7 - 8 | 1235 | 131 | |
GenBank mRNA | CO563709.1 | - | 3 - 4 | 430 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |