Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178258.1 | NP_009535.1 | 1 - 2 | 51 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |