Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180857.3 | NP_010923.3 | 1 | 45 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Amplicon spans exons and probe does not span exons |