Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001180026.1 | NP_011947.2 | 1 - 2 | 117 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Probe spans exons |