Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182080.1 | NP_013294.1 | 1 | 450 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |