Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182187.1 | NP_013402.1 | 1 | 1856 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |