Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001182425.1 | NP_013645.1 | 1 | 948 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |