Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244404.1 | NP_001231333.1 | 5 - 6 | 553 | 92 | |
GenBank mRNA | AK231114.1 | - | 5 - 6 | 529 | 92 | |
AK343771.1 | - | 5 - 6 | 536 | 92 | ||
AK400079.1 | - | 5 - 6 | 550 | 92 | ||
AY609662.1 | - | 5 - 6 | 512 | 92 | ||
FS684590.1 | - | 5 - 6 | 553 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |