Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244707.1 | NP_001231636.1 | 2 - 3 | 271 | 100 | |
XM_005653719.3 | XP_005653776.1 | 299 | 100 | |||
XM_013977625.2 | XP_013833079.1 | 344 | 100 | |||
XM_021102024.1 | XP_020957683.1 | 204 | 100 | |||
GenBank mRNA | AK237186.1 | - | 2 - 3 | 271 | 100 | |
AK395657.1 | - | 2 - 3 | 280 | 100 | ||
AY609450.1 | - | 1 - 2 | 112 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |