Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243720.1 | NP_001230649.1 | 1 - 2 | 234 | 82 | |
XM_013979910.2 | XP_013835364.1 | 291 | 82 | |||
XM_021102005.1 | XP_020957664.1 | 292 | 82 | |||
GenBank mRNA | AK231579.1 | - | 1 - 2 | 234 | 82 | |
AK236842.1 | - | 1 - 2 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |