Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244341.1 | NP_001231270.1 | 1 - 2 | 144 | 79 | |
XM_021093389.1 | XP_020949048.1 | 1101 | 79 | |||
XM_021093390.1 | XP_020949049.1 | 793 | 79 | |||
GenBank mRNA | AK343562.1 | - | 2 - 3 | 1094 | 79 | |
AK395502.1 | - | 2 - 3 | 1082 | 79 | ||
FD613790.1 | - | 1 - 2 | 144 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |