Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925291.5 | XP_001925326.1 | 2505 | 95 | ||
XM_005659599.3 | XP_005659656.1 | 2467 | 95 | |||
XM_005659601.3 | XP_005659658.1 | 2509 | 95 | |||
XM_013992986.2 | XP_013848440.1 | 2455 | 95 | |||
XM_021095334.1 | XP_020950993.1 | 2556 | 95 | |||
XM_021095339.1 | XP_020950998.1 | 2455 | 95 | |||
GenBank mRNA | AF050176.1 | - | 1 | 150 | 95 | |
AK392768.1 | - | 5 | 1152 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |