Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929372.7 | XP_001929407.6 | 1978 | 107 | ||
GenBank mRNA | AJ536287.1 | - | 4 - 5 | 187 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |